0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vacuum Traps
Accessories for Cold Trap for Evaporation Tasks, CT50 Single OLÉ
Accessories for Cold Trap for Evaporation Tasks, CT50 Single OLÉ
Accessories for Cold Trap for Evaporation Tasks, CT50 Single OLÉ
Catalog # HUBE504545
Supplier:
HUBER
New Product
Expand image
Accessories for Cold Trap for Evaporation Tasks, CT50 Single OLÉ
Catalog # HUBE504545
Supplier:
HUBER
New Product
Specifications
Description:
Glass adaptor, 50 mm flange to GL14 for connection to rotary evaporators
Environmentally Preferable:
Specifications
About this item
Product Details
Product Details