ContactUsLinkComponent
ProCare Protein Check
ProCare Protein Check
Catalog # MIEL11626810
Supplier:  MIELE
New Product
CAS Number:  
ProCare Protein Check
Catalog # MIEL11626810
Supplier:  MIELE
New Product
CAS Number:  
Specifications
  • Description:
    Protein check
Specifications
About this item
Protein check  for the detection and qualitative assessment of possible protein residues.
  • Test for residues after cleaning and thermal disinfection
    Precise application
    Short exposure time of 3 minutes
    Three pens per packaging unit
    Up to 50 applications per pen
    Thermodisinfectors
    Cleaning and disinfection machines, medicine
    External dimension: 157×232×28 mm