ContactUsLinkComponent
BradyPrinter i7100 Label Printers with Peel Function and Brady Workstation Suite
BradyPrinter i7100 Label Printers with Peel Function and Brady Workstation Suite
Supplier:  Brady
New Product
Certificates
BradyPrinter i7100 Label Printers with Peel Function and Brady Workstation Suite
Supplier:  Brady
New Product
Specifications
  • Catalog No:
  • BRDY322877
  • BRDY322876
  • BRDY322880
  • BRDY322881
  • Compatibility:
  • Brady workstation, windows based driver for 3rd party software use
  • Brady workstation, windows based driver for 3rd party software use
  • Brady workstation, windows based driver for 3rd party software use
  • Brady workstation, windows based driver for 3rd party software use
  • Connections:
  • Bluetooth (optional), ethernet, I/O connection, serial, USB, Wi-Fi (optional)
  • Bluetooth (optional), ethernet, I/O connection, serial, USB, Wi-Fi (optional)
  • Bluetooth (optional), ethernet, I/O connection, serial, USB, Wi-Fi (optional)
  • Bluetooth (optional), ethernet, I/O connection, serial, USB, Wi-Fi (optional)
  • Display:
  • Colour LCD screen
  • Colour LCD screen
  • Colour LCD screen
  • Colour LCD screen
  • Interfaces:
  • PC, touchscreen
  • PC, touchscreen
  • PC, touchscreen
  • PC, touchscreen
  • Memory:
  • 256 MB RAM, 50 MB IFFS flash, SD card up to 512 GB
  • 256 MB RAM, 50 MB IFFS flash, SD card up to 512 GB
  • 256 MB RAM, 50 MB IFFS flash, SD card up to 512 GB
  • 256 MB RAM, 50 MB IFFS flash, SD card up to 512 GB
  • Model:
  • i7100-300P-UK-BWS
  • i7100-300P-EU-BWS
  • i7100-600P-EU-BWS
  • i7100-600P-UK-BWS
  • Power supply:
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • Print speed:
  • 150 mm/s
  • 150 mm/s
  • 150 mm/s
  • 150 mm/s
  • Print width:
  • 105,6 mm
  • 105,6 mm
  • 105,6 mm
  • 105,6 mm
  • Resolution:
  • 300 dpi
  • 300 dpi
  • 600 dpi
  • 600 dpi
  • Supported OS:
  • Windows 10, Windows 7, Windows 8, Windows server 2012, Windows server 2012 R2, Windows server 2016
  • Windows 10, Windows 7, Windows 8, Windows server 2012, Windows server 2012 R2, Windows server 2016
  • Windows 10, Windows 7, Windows 8, Windows server 2012, Windows server 2012 R2, Windows server 2016
  • Windows 10, Windows 7, Windows 8, Windows server 2012, Windows server 2012 R2, Windows server 2016
  • Temperature range:
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C