ContactUsLinkComponent
High-Resolution Digital Microscope, AM8917MZT4
High-Resolution Digital Microscope, AM8917MZT4
Catalog # IDCPAM8917MZT4
New Product
High-Resolution Digital Microscope, AM8917MZT4
Catalog # IDCPAM8917MZT4
New Product
Specifications
  • Magnification:
    400 ~ 470x
  • Resolution:
    8,0 Megapixel (3840×2160)
Specifications
About this item
AM8917MZT4 features EDOF for expanding the depth of field, offering 8-megapixels of clarity at 400 ~ 470x and incorporates features like DPQ, refocus mode, AMR, eFLC, and even a polariser for assisting professionals in high magnification tasks.
  • White LEDs
    USB 2,0 Interface
    133(H)×33(D) mm dimensions
    Approximately 120 g with 1,8 m cable length
    10 fps at 8,0 MP, MJPEG 25 fps at 2,0 MP, MJPEG maximum frame rate
    With magnification lock, measurement, calibration and microtouch sensor features
    Windows 10/11 macOS 10,14 or later operating system
Certifications:
FCC, ROHS.
Packaging:
Contains microscope, carry pouch, calibration target, user manual, N3C-O- open cap, N3C-C- closed cap, N3C-D- diffuser cap, N3C-E- extension cap, N3C-L- long cap, N3C-S- side light cap.
Caution:
Two years European warranty.