Conductive Tips, Type H
Nome del fornitore: Ritter, part of Avantor
New Product
Certificati
Informazioni su questo articolo
Suitable for use on third party robotic instruments Hamilton Microlab Star and Nimbus (CO-RE version).
Robotic tips in blister packaging are manufactured under cleanroom conditions and tested from independent laboratories to be free of DNA, DNase, RNase and PCR inhibitors. Ritter conductive tips type H are available with or without filter and are CE-IVD marked for clinical diagnostic applications.
With the conductive robotic tips from Ritter Medical, even small volumes can be dosed with high precision and without contamination. The conductivity enables the system to recognize the filling height and will guarantee a minimal immersion of the tip into the liquid, in order to assure a save pipetting and dispensing
Frequently Bought Together