Stainless Steel Heavy Duty Tables with Perforated Top
: TEKNOMEK
New Product
The stainless steel rear tie bar heavy duty table with perforated top is an ideal surface for industries that require sterile, hygienic and contamination resistant surfaces. It is more sturdy with a heavier payload for items that are of a heavier weight.
Frame in 304 L-grade stainless steel
Brushed dull polish finish
304-grade perforated top with a silicone seal
304-grade perforated undershelf
4× stainless steel feet with non-marking floor levelling rubber base
Payload of up to 250 kg
Floor clearance to undershelf: 155 mm
With both the top and optional shelf perforated for optimum airflow and an all 304 L-grade stainless steel construction, it's the perfect hygienic work surface for any clean environment.
Frequently Bought Together