ContactUsLinkComponent
Tegaderm™ Antimicrobial I.V. Advanced Securement Dressing
Tegaderm™ Antimicrobial I.V. Advanced Securement Dressing
Catalog # MMME9132
Supplier:  3M MEDICAL
New Product
Tegaderm™ Antimicrobial I.V. Advanced Securement Dressing
Catalog # MMME9132
Supplier:  3M MEDICAL
New Product
Specifications
  • Description:
    Antimicrobial I.V. advanced securement dressing, without latex
  • L×W:
    85×70 mm
Specifications
About this item
The Tegaderm™ Antimicrobial I.V. Advanced Securement Dressing is a peripheral IV securement dressing with chlorhexidine gluconate (CHG) formulated into the dressing adhesive. Its integrated design combines antimicrobial protection with site visibility, catheter securement and consistent application for peripheral IVs.
  • Designed to minimise catheter movement and dislodgement
    Breathable; semi-permeable
    Comfortable to wear and conformable, permits free movement
    Large securement tape strip with notch promotes consistent, correct application
    Transparent film allows continuous site visibility toeasily assess for early signs of infection
    Waterproof, sterile barrier protects against external contaminants
Antimicrobial IV securement dressing with 2% CHG formulated into the adhesive offers antimicrobial protection without requiring additional moisture to activate.
Integrated design combines antimicrobial protection with site visibility, catheter securement and consistent application. Proven to suppress regrowth of skin flora for up to seven days for sustained antimicrobial protection.