ContactUsLinkComponent
Tork Surface Disinfectant Sprays
Tork Surface Disinfectant Sprays
Catalog # SCAA193610
Supplier:  Essity
New Product
undefined
Tork Surface Disinfectant Sprays
Catalog # SCAA193610
Supplier:  Essity
New Product
Specifications
  • Description:
    Surface disinfecting spray, refill, premium
  • Environmentally Preferable:
  • Packed:
    6 bottles per box, 500 ml each (6×500 ml)
Specifications
About this item
The surface disinfecting spray is suitable for PT2 (disinfectants not intended for direct application to humans or animals) and PT4 (food and feed area) applications, food contact surfaces must be rinsed with drinking water after disinfecting with product. The product is an alcohol-free ready-to-use multi-purpose spray for surface disinfection.
  • Kills 99,99% of enveloped viruses in five minutes including coronavirus EN14476: Limited virucidal activity (Active against enveloped viruses such as, e.g. Influenza virus, coronavirus, HIV, herpes virus, hepatitis virus) exposure time: 5 minutes under dirty conditions
    Biodegradable formula
    100% plant-based active ingredient (Lactic acid)
    Food surface contact safe
    No scent fragrance
    Premium quality
    Colour: Transparent
Tork surface disinfecting spray is a ready-to-use disinfectant that is effective against a broad spectrum of bacteria, viruses and yeasts, including Coronavirus. It is a biodegradable formula and contains 100% plant-based active ingredient (lactic acid). The surface disinfecting spray is food surface contact safe. The product is alcohol-free and is available in a 500 ml. spray bottle with a dual function trigger spray nozzle (both spray and foam).