ContactUsLinkComponent
Millipore® Immobilon®-FL NOW Bundle Hydrophobic PVDF Transfer Membrane
Millipore® Immobilon®-FL NOW Bundle Hydrophobic PVDF Transfer Membrane
  MILFNWKITFL
 :  Merck
New Product
undefined
Millipore® Immobilon®-FL NOW Bundle Hydrophobic PVDF Transfer Membrane
  MILFNWKITFL
 :  Merck
New Product
  • Description:
    Immobilon®-FL NOW bundle transfer membrane
  • Dimensions:
    85×10000 mm
Hydrophobic PVDF transfer membrane for western blotting using fluorescent, chemifluorescence, chromogenic, or chemiluminescence.
  • Western blotting
    Hydrophobic
    Plain/white filter
    0,45 µm pore size
    Comes in a 8,5 cm × 10 m roll
The Immobilon NOW system offers 1-cut convenience for standard mini-blots (7 × 8,5cm) and midi-blots (8,5 × 13,5cm).
Compatible stains: All fluorescence dyes, Coomassie brilliant blue, Amido black, Ponceau-S red and Sypro® ruby