ContactUsLinkComponent
Millipore® Immobilon®-PSQ NOW Bundle Hydrophobic PVDF Transfer Membrane
Millipore® Immobilon®-PSQ NOW Bundle Hydrophobic PVDF Transfer Membrane
Catalog # MILFNWKITSQ
Supplier:  Merck
New Product
undefined
Millipore® Immobilon®-PSQ NOW Bundle Hydrophobic PVDF Transfer Membrane
Catalog # MILFNWKITSQ
Supplier:  Merck
New Product
Specifications
  • Description:
    Immobilon®-PSQ NOW bundle hydrophobic PVDF transfer membrane
  • Dimensions:
    265×3750 mm
Specifications
About this item
PVDF transfer membrane hydrophobic for western blotting using chemiluminescence, chromogenic or radioactive detection methods.
  • Western blotting
    Dot blot
    White/plain filter
    Electrostatic, hydrophobic
    0,2 µm filter pore size
    Comes in an 8,5 cm × 10 m roll
The Immobilon NOW system offers 1-cut convenience for standard mini-blots (7×8,5 cm) and midi-blots (8,5×13,5 cm).
Compatible stainn: Coomassie brilliant blue, Amido black, India ink, Ponceau-S red, Colloidal gold, CPTS, Toluidine blue, Transillumination and Sypro® ruby.