Orders LinkContactUsLinkComponent
CytoFLEX Mosaic Detection Modules
CytoFLEX Mosaic Detection Modules
New Product
Supplier:   Beckman Coulter
CytoFLEX Mosaic Detection Modules
Supplier:  Beckman Coulter
New Product
Certificates
CytoFLEX Mosaic Detection Modules
CytoFLEX Mosaic Detection Modules
14 Options Available Below
Product Details & Documents
The CytoFLEX Mosaic Detection Module is the natural evolution of the CytoFLEX platform. A smart approach to spectral analysis, the detection module connects to the CytoFLEX LX/S flow cytometer, providing the flexibility to mix and match different detection methods in one instrument.
  • A flexible approach to spectral flow cytometry
With up to 88 detection channels, the CytoFLEX Mosaic Detection Module enhances the CytoFLEX S or LX flow cytometer, empowering users to tackle the most complex experiments. Together, they create the most advanced and powerful CytoFLEX flow cytometry solution to date.
Product Family Options
Product Information
  • Availability
    Price
  • BECMD19581
    CytoFLEX Mosaic 88 spectral detection module U-V-B-Y-R-I (6 laser CytoFLEX LX UV)
    CytoFLEX Mosaic 88 spectral detection module U-V-B-Y-R-I (6 laser CytoFLEX LX UV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD19582
    CytoFLEX Mosaic 88 spectral detection module U-V-B-Y-R (5 laser CytoFLEX LX UV)
    CytoFLEX Mosaic 88 spectral detection module U-V-B-Y-R (5 laser CytoFLEX LX UV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD19583
    CytoFLEX Mosaic 88 spectral detection module U-V-B-R (4 laser CytoFLEX LX UV)
    CytoFLEX Mosaic 88 spectral detection module U-V-B-R (4 laser CytoFLEX LX UV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD19584
    CytoFLEX Mosaic 88 spectral detection module N-V-B-Y-R-I (6 laser CytoFLEX LX NUV)
    CytoFLEX Mosaic 88 spectral detection module N-V-B-Y-R-I (6 laser CytoFLEX LX NUV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD19585
    CytoFLEX Mosaic 88 spectral detection module N-V-B-Y-R (5 laser CytoFLEX LX NUV)
    CytoFLEX Mosaic 88 spectral detection module N-V-B-Y-R (5 laser CytoFLEX LX NUV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD19586
    CytoFLEX Mosaic 88 spectral detection module V-B-Y-R (4 laser CytoFLEX LX NUV)
    CytoFLEX Mosaic 88 spectral detection module V-B-Y-R (4 laser CytoFLEX LX NUV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD20160
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-Y-R-I (6 laser UV)
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-Y-R-I (6 laser UV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD20161
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-Y-R (5 laser UV)
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-Y-R (5 laser UV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD20162
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-R (4 laser UV)
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-R (4 laser UV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD20163
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system N-V-B-Y-R-I (6 laser NUV)
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system N-V-B-Y-R-I (6 laser NUV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD20164
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system N-V-B-Y-R (5 laser NUV)
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system N-V-B-Y-R (5 laser NUV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD20165
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system V-B-Y-R (4 laser NUV)
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system V-B-Y-R (4 laser NUV)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD20166
    CytoFLEX S and CytoFLEX Mosaic 63 spectral flow analyzer system B-R-V-Y (4 laser 561)
    CytoFLEX S and CytoFLEX Mosaic 63 spectral flow analyzer system B-R-V-Y (4 laser 561)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • BECMD19589
    CytoFLEX Mosaic computer and monitor
    CytoFLEX Mosaic computer and monitor
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4’841.00
    SFr. 4’841.00
    1 * 1 ST
       SFr. 4’841.00
    Specifications:
    New Product
    New Product
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..