Order Entry
Switzerland
ContactUsLinkComponent
NANOCOLOR® Photometric Standard Test Kit for Chlorine
NANOCOLOR® Photometric Standard Test Kit for Chlorine
Catalog # MANA91816
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Standard Test Kit for Chlorine
Catalog # MANA91816
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    DPD
  • Pk:
    250 Tests
  • Description:
    NANOCOLOR® Chlorine, test no. 1-16
  • Dimensions:
    160×110×130 mm (Including packaging)
  • Platform:
    NANOCOLOR® standard tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    36
  • Weight:
    255,2 g (Including packaging)
  • For unit:
    UV/VIS II; VIS II; Advance; 500 D; MT Easy VIS; MT Easy UV
  • Tests per kit:
    250
  • Range:
    0,02 - 10,0 mg/L (Cl₂); 0,02 - 10,0 mg/L (Cl₂)

Specifications

About this item

Photometric test kit for the determination of chlorine in water samples.

  • Ready-to-use reagents for up to 500 determinations
  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: Yes
Hazardous material: No
Remark: Measuring range on NANOCOLOR VIS II

Sludge reagent set: No

Digestion set: No

NanOx Metal: No

NanOx N: No

Delivery: Rugged box with reagents for 250 tests.