NANOCOLOR® Photometric Standard Test Kit for Ozone
Catalog # MANA91885
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:Indigotrisulfonate
- Pk:1 SET
- Description:NANOCOLOR® Ozone, test no. 1-85
- Dimensions:160×110×130 mm (Including packaging)
- Platform:NANOCOLOR® standard tests
- Storage temperature:2...8 °C
- Shelf life (months):12
- Weight:534,4 g (Including packaging)
- For unit:UV/VIS II; VIS II; Advance; 500 D
- Tests per kit:200
- Range:0,01 - 1,50 mg/L (O₃); 0,01 - 1,50 mg/L (O₃)
Specifications
About this item
Photometric test kit for the determination of ozone in water samples.
Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Delivery:
Rugged box with reagents for 200 tests.