Order Entry
Switzerland
ContactUsLinkComponent
NANOCOLOR® Photometric Standard Test Kit for Aluminium
NANOCOLOR® Photometric Standard Test Kit for Aluminium
Catalog # MANA91802
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Standard Test Kit for Aluminium
Catalog # MANA91802
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Eriochrome Cyanine R
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Aluminum, test no.1-02
  • Dimensions:
    160×110×130 mm (including packaging)
  • Seq:
    0
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    24
  • Weight:
    616 g (including packaging)
  • Type:
    Hazardous material: No
  • For unit:
    UV/VIS II; VIS II; Advance; 500 D; MT Easy VIS; MT Easy UV
  • Tests per kit:
    250
  • Range:
    0,01 - 1,00 mg/L (Al³⁺)

Specifications

About this item

Photometric test kit for the determination of aluminium in water samples.

  • Ready-to-use reagents for up to 500 determinations

  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: Yes
Hazardous material: No

Remark: Measuring range on NANOCOLOR VIS II

Delivery: Remark: Measuring range on NANOCOLOR VIS II