NANOCOLOR® Photometric Standard Test Kit for Zinc
MANA91895
:
- Chemical method:Zincon
- Pk:1 KIT
- Description:NANOCOLOR® Zink, test no. 1-95
- Dimensions:160×110×130 mm (Including packaging)
- Model:MANA91895
- Platform:NANOCOLOR® standard tests
- Storage temperature:15...25 °C
- Shelf life (months):36
- Weight:464,7 g (Including packaging)
- For unit:UV/VIS II;VIS II;Advance;500 D;MT Easy VIS;MT Easy UV
- Tests per kit:250
- Range:0,02 - 3,0 mg/L (Zn²⁺)
Photometric test kit for the determination of zinc in water samples.
- Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
: Rugged box with reagents for 250 tests.