NANOCOLOR® Photometric Standard Test Kit for Lead
N° de catalogue MANA918101
Fournisseur: MACHEREY-NAGEL
Numéro CAS:
Spécifications
- Chemical method:Dithizone
- Pk:1 KIT
- Description:NANOCOLOR® Lead, test no. 1-101, not for sea water analysis
- Dimensions:160×110×255 mm (including packaging)
- Model:MANA918101
- Platform:NANOCOLOR® standard tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:750 g (including packaging)
- For unit:UV/VIS II;VIS II;Advance;500 D
- Tests per kit:25
- Range:0,005 - 1,00 mg/L (Pb²⁺)
Spécifications
À propos de cet article
Photometric test kit for the determination of lead in water samples.
Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Livraison:
Two rugged boxes with reagents for 25 tests.