Order Entry
Switzerland
ContactUsLinkComponent
Scepter™ 2.0 Automated Cell Counter
Scepter™ 2.0 Automated Cell Counter
Catalog # MILXPHCC20060
Supplier:  Merck
Scepter™ 2.0 Automated Cell Counter
Catalog # MILXPHCC20060
Supplier:  Merck

Specifications

  • Description:
    Scepter 2.0 Cell counter with 60 μm Scepter sensors (50 pack)
  • Seq:
    2
  • Environmentally Preferable:

Specifications

About this item

The Scepter cell counter accurately count particles smaller than 6 µm in diameter.

  • Easy-to-use, portable format

    Counts blood cells and other cells with small diameters with the highest precision

The 40 µm sensor can count samples concentrated up to 1500000 cells/ml, 60 µm sensor can count up to 500000 cells/ml.

Powerful software for complex, effortless cell analysis, to compare sample sets side by side using histogram overlay and multiparametric tables, save and create gating templates, generate reports, graphs and tables.