Order Entry
Switzerland
ContactUsLinkComponent
WHEATON® Lyophilisation Stopper, Bromobutyl
WHEATON® Lyophilisation Stopper, Bromobutyl
Catalog # LSUK700004
Supplier:  DWK Life Sciences
undefined
WHEATON® Lyophilisation Stopper, Bromobutyl
Catalog # LSUK700004
Supplier:  DWK Life Sciences

Specifications

  • Colour:
    Grey
  • Description:
    Bromobutyl lyophilisation stopper, 13 mm

Specifications

About this item

Igloo lyophilisation style stoppers.

  • Manufactured from the highest quality bromobutyl rubber formulation

  • Components manufactured to yield exceptional dimensional stability

Food safe (EU only).