WHEATON® Plastic Media Bottle, PC
Catalog # WHEAW017795
Supplier: DWK Life Sciences
Specifications
- Cap Size:53B
- Capacity:2000 ml
- Colour:Clear
- Material:PC
Specifications
About this item
These WHEATON® square/octagonal plastic media bottles are manufactured from polycarbonate with blue tamper-evident linerless polypropylene cap attached.
Ideal for the storage and transport of media, laboratory reagents, and biological buffers
- Molded graduations provide a guide for volume filling
- Cleanroom packaged with multi-barrier elastomer packaging
- Square footprint ensures efficient use of storage space
- Autoclavable
These polycarbonate bottles are supplied non-sterile, but can be safely sterilized by gamma irradiation without property loss or color change.
WHEATON® provides customers the option of personalizing their container orders by adding a company logo of their choice to the product cap.