TriLink® N1-Methoxymethylpseudouridine-5'-Triphosphate
Supplier: TriLink BioTechnologies
New Product
Certificates
About this item
- Grade: RUO
Caution: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
Specifications
- Application:
- Environmentally Preferable:
Specifications
Frequently Bought Together