TriLink® N6-Methyladenosine-5'-Triphosphate
New Product
N6-methyl adenosine (N6-methyl ATP) is a base modified analog of adenosine and is found as a minor nucleoside in natural RNAs (Meyer et al.). N6-methyl ATP can substitute for ATP in some biological systems, and is a potent agonist for P2Y-purinoceptors in the guinea pig, taenia coli (Burnstock et al.).
- Grade: RUO
: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
Frequently Bought Together