Orders LinkContactUsLinkComponent
b.safe Disc Sensor Holders
b.safe Disc Sensor Holders
Supplier:   Bohlender
b.safe Disc Sensor Holders
Supplier:  Bohlender
Certificates
b.safe Disc Sensor Holders
b.safe Disc Sensor Holders
2 Options Available Below
Product Details & Documents
Made of PE, height-adjustable bracket for a b.safe disc sensor (catalogue number BOHLR640-05).
  • Simple attachment of the b.safe disc sensor by hanging the holder into the b.safe waste cap
  • Position of the sensor can be stepless adjusted in height for level monitoring
  • Use in containers and canisters made of non-conductive materials
The disc sensor is clipped into the corresponding holder, the cable guide prevents kinks on the sensor cable. After having put the holder on the PTFE insert of the waste cap, the disc sensor can be positioned freely at the required height. The adjusted height corresponds to the fill level to start the alarm.
Product Family Options
Product Information
  • For
    Availability
    Price
  • BOHLR649-45
    Disc sensor holder, 81×130 mm
    Disc sensor holder, 81×130 mm
    Waste cap, GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 135.00
    SFr. 135.00
    1 * 1 ST
       SFr. 135.00
  • BOHLR649-55
    Disc sensor holder, 91×130 mm
    Disc sensor holder, 91×130 mm
    Waste cap, S55
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 156.00
    SFr. 156.00
    1 * 1 ST
       SFr. 156.00