Order Entry
Switzerland
ContactUsLinkComponent
SUPL1047321
SUPL1047321
Catalog # 1047321000.
Supplier:  Merck
New Product
   Safer Human and Environmental Health
CAS Number:  
undefined
SUPL1047321
Catalog # 1047321000.
Supplier:  Merck
New Product
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Borosilicate glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • EPA 533/537.1 Tested:
    Conforms
  • UHPLC Gradient (at 210 nm):
    ≤1.0 mAU
  • Evaporation Residue:
    ≤0.7 mg/l
  • Titratable Acid (meq/g):
    ≤0.001
  • Titratable Base (meq/g):
    ≤0.0001

Specification Test Results

About this item

 :
 :32.04 g/mol
UN:1230
REACH:01-2119433307-44-XXXX
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Room Temperature
CAS Number:67-56-1
Formula:CH₄O
MDL Number:MFCD00004595