Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Staphylococcus aureus Recombinant Protein A (from E. coli)

Staphylococcus aureus Recombinant Protein A (from E. coli)

Catalog # ABCAAB52953-10
Marketsource
Supplier:   Abcam
Staphylococcus aureus Recombinant Protein A (from E. coli)
Catalog # ABCAAB52953-10
Supplier:  Abcam
Marketsource
CAS Number:  
Staphylococcus aureus Recombinant Protein A (from E. coli)
Staphylococcus aureus Recombinant Protein A (from E. coli)
CAS Number:  
Price
SFr. 200.00
Log in for availability
1 * 10 mg
Product Details & Documents

Recombinant Staphylococcus aureus Protein A is a Staphylococcus aureus subsp. aureus NCTC 8325 Fragment protein, in the 32 to 327 aa range, expressed in E. coli, with >99% purity, <0,1 EU/mg endotoxin level and suitable for SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    Protein A
  • Protein synonyms:
    SAOUHSC_00069|spa|Immunoglobulin G-binding protein A|IgG-binding protein A|Staphylococcal protein A|SpA
  • Purity:
    >99%
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB52953-10
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0,1 EU/mg
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    Constituents: Water.
  • Conjugation:
    Unconjugated
  • Tested applications:
    SDS-PAGE
  • Species:
    Staphylococcus aureus
  • Animal-free:
    Y

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.