Human Recombinant SEC13L1 (from E. coli)
Recombinant Human SEC13L1 protein is a Human Full Length protein, in the 1 to 322 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Form:Liquid
- Protein/peptide name:SEC13L1
- Protein synonyms:D3S1231E|SEC13A|SEC13L1|SEC13R|SEC13|Protein SEC13 homolog|GATOR2 complex protein SEC13|SEC13-like protein 1|SEC13-related protein
- Purity:>95%
- Concentration:0,5 mg/ml
- Source:E. coli
- Biological activity:Standard
- Shipping temperature:20
- Cat. No.:ABCAAB105635-100
- Protein/peptide type:Recombinant
- Storage conditions:Store at −20 °C
- Formulation:pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Conjugation:Unconjugated
- Tested applications:MS, SDS-PAGE
- Species:Human
- Animal-free:N
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



