Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Human Recombinant PMS2 (from Wheat germ)

Human Recombinant PMS2 (from Wheat germ)

Catalog # ABCAAB131836-10
Marketsource
Supplier:   Abcam
Human Recombinant PMS2 (from Wheat germ)
Catalog # ABCAAB131836-10
Supplier:  Abcam
Marketsource
CAS Number:  
Human Recombinant PMS2 (from Wheat germ)
Human Recombinant PMS2 (from Wheat germ)
CAS Number:  
Price
SFr. 714.00
Log in for availability
1 * 10 µG
Product Details & Documents

Recombinant Human PMS2 protein is a Human Full Length protein, in the 1 to 862 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    PMS2
  • Protein synonyms:
    PMSL2|PMS2|Mismatch repair endonuclease PMS2|DNA mismatch repair protein PMS2|PMS1 protein homolog 2
  • Source:
    Wheat germ
  • Biological activity:
    Standard
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCAAB131836-10
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at –80 °C
  • Formulation:
    pH: 8 Constituents: 0,79% Tris HCl, 0,31% Glutathione
  • Conjugation:
    Unconjugated
  • Tested applications:
    ELISA, SDS-PAGE, WB
  • Species:
    Human
  • Animal-free:
    N

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.