Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Human SSH2 ELISA Kits

Human SSH2 ELISA Kits

New Product
Supplier:   Antibodies.com
Human SSH2 ELISA Kits
Supplier:  Antibodies.com
New Product
Certificates

2 Options Available Below

Product Details & Documents

Human SSH2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SSH2 in tissue homogenates or other biological fluids.

Human SSH2 ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SSH2 in tissue homogenates or other biological fluids. An antibody specific for SSH2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SSH2 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-SSH2 Antibody, which binds the captured SSH2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of SSH2 captured in each well. The concentration of SSH2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ANTIA7360-48
  • ANTIA7360-96
  • Target protein:
  • SSH2
  • SSH2
  • Primary antibody reactivity:
  • Human
  • Human
  • Assay Type:
  • Sandwich
  • Sandwich
  • Format:
  • Pre-coated
  • Pre-coated
  • Assay duration:
  • Multiple steps
  • Multiple steps
  • Sensitivity:
  • 0,113 ng/ml
  • 0,113 ng/ml
  • Detection range:
  • 0,312 - 20 ng/ml
  • 0,312 - 20 ng/ml
  • Detection method:
  • Colorimetric
  • Colorimetric
  • Detection limit:
  • Intra-assay: CV <10%, Inter-assay: CV <12%
  • Intra-assay: CV <10%, Inter-assay: CV <12%
  • Sample type:
  • Tissue homogenates or other biological fluids
  • Tissue homogenates or other biological fluids
  • Time to Results:
  • 4 h 30 min
  • 4 h 30 min
  • Regulatory status:
  • RUO
  • RUO
  • Storage temperature:
  • 4 °C
  • 4 °C
  • Shelf life:
  • Store for 6 months at 4 °C
  • Store for 6 months at 4 °C

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • ANTIA7360-48
    Human SSH2 ELISA kit
    Human SSH2 ELISA kit
    Log in for availability
    Log in for availability
    1 * 48 Tests
    1 * 48 Tests - SFr. 604.00
    SFr. 604.00
    1 * 48 Tests
       SFr. 604.00

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ANTIA7360-48
    Target protein
    SSH2
    Primary antibody reactivity
    Human
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    Multiple steps
    Sensitivity
    0,113 ng/ml
    Detection range
    0,312 - 20 ng/ml
    Detection method
    Colorimetric
    Detection limit
    Intra-assay: CV <10%, Inter-assay: CV <12%
    Sample type
    Tissue homogenates or other biological fluids
    Time to Results
    4 h 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
    Shelf life
    Store for 6 months at 4 °C
    New Product
  • ANTIA7360-96
    Human SSH2 ELISA kit
    Human SSH2 ELISA kit
    Log in for availability
    Log in for availability
    1 * 96 Tests
    1 * 96 Tests - SFr. 794.00
    SFr. 794.00
    1 * 96 Tests
       SFr. 794.00

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ANTIA7360-96
    Target protein
    SSH2
    Primary antibody reactivity
    Human
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    Multiple steps
    Sensitivity
    0,113 ng/ml
    Detection range
    0,312 - 20 ng/ml
    Detection method
    Colorimetric
    Detection limit
    Intra-assay: CV <10%, Inter-assay: CV <12%
    Sample type
    Tissue homogenates or other biological fluids
    Time to Results
    4 h 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
    Shelf life
    Store for 6 months at 4 °C
    New Product