Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Blocking Buffer 10X

Blocking Buffer 10X

Blocking buffer, uncoated, 10X, 50 ml
Catalog # ABCAAB210904-50
Marketsource
Supplier:   Abcam
Blocking Buffer 10X
Catalog # ABCAAB210904-50
Supplier:  Abcam
Marketsource
CAS Number:  
Blocking Buffer 10X
Blocking Buffer 10X

Blocking buffer, uncoated, 10X, 50 ml

CAS Number:  
Price
SFr. 170.00
Log in for availability
1 * 50 mL
Product Details & Documents

Blocking Buffer 10X is part of the reagents, controls and accessories range.

 : RUO.

Specifications

Specifications

  • Concentration:
    10X
  • Description:
    Blocking buffer, uncoated
  • Volume:
    50 ml
  • Cat. No.:
    ABCAAB210904-50
  • Storage temperature:
    4 °C

Specifications