Order Entry
Switzerland
ContactUsLinkComponent
Ciprofloxacin ELISA Kit
Ciprofloxacin ELISA Kit
Catalog # ABCAAB285266-96
Supplier:  Abcam
CAS Number:  
undefined
Ciprofloxacin ELISA Kit
Catalog # ABCAAB285266-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Competitive
  • Description:
    Ciprofloxacin (Cipro) ELISA Kit, 100 assays

Specifications

About this item

A competitive ELISA kit for quantify of Ciprofloxacin in tissue, honey, eggs, milk, milk powder.

  • Easy, convenient and time-saving method to measure the level of Ciprofloxacin in tissue, honey, eggs, milk, milk powder
  • Detection Range: 0,5 to 40 ppb (ng/ml)
  • Sensitivity: <0,05 ppb
  • Detection limit: 0,3 ppb for tissue, 0,4 ppb for honey, 3 ppb for milk and egg, 6 ppb for milk powder
  • Cross Reactivity: Sarafloxacin −110%; Oxolin acid −28%; Levofloxacin −10%; Lomefloxacin, Marbofloxacin −4%

Ciprofloxacin is an antibiotic used to treat a number of bacterial infections. It is commonly used to treat acne and rosacea. Historically it was important in reducing the number of deaths from cholera. A broad-spectrum antibiotic of the polyketide class, it is produced by the actinobacterial genus Streptomyces. It acts by inhibiting protein synthesis. It is first-line therapy for rocky mountain spotted fever, Lyme disease, Q fever, psittacosis and lymphogranuloma venereum, mycoplasma pneumoniae and to eradicate nasal carriage of meningococci. BioVision’s Ciprofloxacin ELISA kit is a competitive ELISA assay for the quantitative measurement of Ciprofloxacin in tissues, honey and urine. The density of colour is proportional to the amount of tetracycline captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.