10X Wash Buffer PT
10X Wash buffer PT, 1000 ml
ABCAAB206977-1000
Marketsource
: Abcam
10X Wash Buffer PT (ab206977) is part of the reagents, controls and accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.
: RUO
- Description:10X Wash buffer PT
- Volume:1000 ml
- Cat. No.:ABCAAB206977-1000
- Form:Pre-coated
- Storage temperature:4 °C



