Human Transferrin ELISA Kit
About this item
Human Transferrin ELISA kit is a single-wash 90-min Simplestep used to quantify Human Transferrin with a sensitivity of 4,396 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.
2 Options Available Below
Product Details & Documents
Human Transferrin ELISA kit is a single-wash 90-min Simplestep used to quantify Human Transferrin with a sensitivity of 4,396 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ABCAAB288174-1X96
- ABCAAB288174-1X384
- Target protein:
- Transferrin
- Transferrin
- Primary antibody reactivity:
- Human
- Human
- Assay Type:
- Sandwich
- Sandwich
- Format:
- Pre-coated
- Pre-coated
- Assay duration:
- One step
- One step
- Sensitivity:
- 4,396 pg/ml
- 4,396 pg/ml
- Detection range:
- 15,625 - 1000 pg/ml
- 15,625 - 1000 pg/ml
- Detection method:
- Colorimetric
- Colorimetric
- Sample type:
- Cell culture media, Heparin Plasma, Citrate plasma, Serum, EDTA Plasma
- Cell culture media, Heparin Plasma, Citrate plasma, Serum, EDTA Plasma
- Time to Results:
- 1 h 30 min
- 1 h 30 min
- Regulatory status:
- RUO
- RUO
- Storage temperature:
- 4 ℃
- 4 ℃
Specifications
Product Family Options
Product Information
- Tests per kitAvailabilityPrice
- ABCAAB288174-1X96Human Transferrin ELISA kitSpecifications:
More
Specifications
Cat. No.ABCAAB288174-1X96Target proteinTransferrinPrimary antibody reactivityHumanAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity4,396 pg/mlDetection range15,625 - 1000 pg/mlDetection methodColorimetricSample typeCell culture media, Heparin Plasma, Citrate plasma, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOStorage temperature4 ℃ - ABCAAB288174-1X384Human Transferrin ELISA kitSpecifications:New Product
More
Specifications
Cat. No.ABCAAB288174-1X384Target proteinTransferrinPrimary antibody reactivityHumanAssay TypeSandwichFormatPre-coatedAssay durationOne stepSensitivity4,396 pg/mlDetection range15,625 - 1000 pg/mlDetection methodColorimetricSample typeCell culture media, Heparin Plasma, Citrate plasma, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOStorage temperature4 ℃New Product



