Anti-SARS-CoV-2 Spike Glycoprotein S1 Human Monoclonal Antibody [clone: CR3022]
ABCAAB273073-200
: Abcam
- Size:200 µg
- Pack type:Vial
Human monoclonal [CR3022] to SARS-CoV-2 Spike Glycoprotein S1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: SARS-CoV-2 Spike Glycoprotein S1
Clonality: Monoclonal
Clone: CR3022
Conjugation:
Epitope: This antibody binds the amino acids 318-510 in the S1 domain of the SARS-CoV Spike protein as well as SARS-CoV-2 (COVID-19) Spike protein. The antibody also binds to P462L-substituted S318–510 fragments of the SARS spike protein. The binding epitope is only accessible in the "open" conformation of the spike protein (Joyce et al. 2020).
Host: Human
Isotype: IgG1
Reactivity: