Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:IL-1 beta
- Description:Rat IL-1 beta ELISA kit (high sensitivity)
- Sample type:Serum, plasma, and other biological fluids
- Cross reactivity:No significant cross-reactivity or interference between rat IL-1 beta and analogues was observed.
- Detection method:Colorimetric
- Time to Results:3 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,31 - 20 pg/ml
- Storage temperature:4 °C
- Sensitivity:0,2 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Rat IL-1 beta ELISA kit is a high sensitivity sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat IL-1 beta in serum, plasma, and other biological fluids.
- High Sensitivity: Detection at the picogram level!
- Assay Precision: Intra-Assay: CV <10%, Inter-Assay: CV <10%
Rat IL-1 beta ELISA kit employs the sandwich Enzyme-Linked Immunoassay technique for the quantitative measurement of rat IL-1 beta in serum, plasma, and other biological fluids. An antibody specific for IL-1 beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-1 beta present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IL-1 beta Antibody, which binds the captured IL-1 beta present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IL-1 beta captured in each well. The concentration of IL-1 beta can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.