Order Entry
Switzerland
ContactUsLinkComponent
Mouse Anti-NMDAR Antibody ELISA Kit
Mouse Anti-NMDAR Antibody ELISA Kit
Catalog # ANTIA319449-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
Mouse Anti-NMDAR Antibody ELISA Kit
Catalog # ANTIA319449-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Anti-NMDAR Antibody
  • Description:
    Mouse Anti-NMDAR Antibody ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Mouse Anti-NMDAR Antibody ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Anti-NMDAR Antibody. No significant cross-reactivity or interference between Anti-NMDAR Antibody and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 hours
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1,563 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,938 ng/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse Anti-NMDAR Antibody ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Anti-NMDAR Antibody in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Delivery: This is a Ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch and protocol manual.
Caution: This product is for Research use only. It is not for use in diagnostic or therapeutic procedures.