Monkey Rheumatoid Factor IgM ELISA Kit
ANTIA319488-96
New Product
:
- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Monkey
- Target protein:Rheumatoid Factor IgM
- Description:Monkey Rheumatoid factor IgM ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection method:Colorimetric
- Time to Results:4 hours
- Shelf life:Store for 6 months at 4 °C
- Detection range:4,688 - 300 IU/ml
- Storage temperature:4 °C
- Sensitivity:2,813 IU/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Monkey Rheumatoid Factor IgM ELISA kit is an indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of monkey Rheumatoid Factor IgM in serum, plasma, tissue homogenates, and other biological fluids.
- Assay Precision: Intra-assay CV: <8%, Inter-assay CV: <10%
- Assay Time: 4 hours
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.