Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Porcine Complement C5a ELISA Kit

Porcine Complement C5a ELISA Kit

Porcine Complement C5a ELISA kit, 96 tests
Catalog # ANTIA319375-96
Supplier:   Antibodies.com
Porcine Complement C5a ELISA Kit
Catalog # ANTIA319375-96
Supplier:  Antibodies.com
CAS Number:  
Porcine Complement C5a ELISA Kit

Porcine Complement C5a ELISA kit, 96 tests

CAS Number:  
Price
SFr. 818.00
Log in for availability
1 * 96 Tests
Product Details & Documents

Porcine Complement C5a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine Complement C5a in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Porcine Complement C5a ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of porcine Complement C5a in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C5a has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C5a present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Complement C5a Antibody, which binds the captured Complement C5a present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C5a captured in each well. The concentration of Complement C5a can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Delivery information: This is a Ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch and protocol manual.

Caution: This product is for Research use only. It is not for use in diagnostic or therapeutic procedures.

Specifications

Specifications

  • Description:
    Porcine Complement C5a ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Other
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    78,125 - 5000 pg/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 hours
  • Cross reactivity:
    Porcine Complement C5a ELISA Kit exhibits high specificity and excellent specificity for the detection of porcine Complement C5a. No significant cross-reactivity or interference between Complement C5a and analogues was observed.
  • Cat. No.:
    ANTIA319375-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    46,875 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Complement C5a
  • Regulatory status:
    RUO

Specifications