Bottle-top dispenser, Calibrex™
Anbieter: Socorex Isba
Zertifikate
Über diesen Artikel
With fix or adjustable volume.
- PFA coated plungerwith broad chemical resistance
- QR coded chemical compatibility
- Integrated calibration key
- Quick disassembling, no tool required
- Autoclavable at 121 °C fully assembled
This model is ranging 1 to 10 ml. The Calibrex™ combine a clever simple construction and a universal chemical compatibility. Appreciated for its smooth, effort-less activation and sturdiness, the base thread fits 32 mm bottle neck diameter.
Delivery: Supplied with three PP adapters for 28, 40 and 45 mm bottle neck diameters - See bottle-neck adapters for additional sizes and materials - and a tube.
Frequently Bought Together