Orders LinkContactUsLinkComponent
Linen testers
Linen testers
Supplier:   ESCHENBACH OPTIK
Linen testers
Supplier:  ESCHENBACH OPTIK
Certificates
Linen testers
4 Options Available Below
Product Details & Documents
Satin matt chrome plated brass frame with silicate glass lens.
Product Family Options
Product Information
  • Magnification
    Ø
    W×D
    Availability
    Price
  • 17,6 mm
    20×20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 78.40
    SFr. 78.40
    1 * 1 ST
       SFr. 78.40
  • 10×
    14,6 mm
    15×15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 78.40
    SFr. 78.40
    1 * 1 ST
       SFr. 78.40
  • 31,6 mm
    30×30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 102.00
    SFr. 102.00
    1 * 1 ST
       SFr. 102.00
  • 22,6 mm
    25×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 78.40
    SFr. 78.40
    1 * 1 ST
       SFr. 78.40