Torches
Anbieter: VARIAN
Zertifikate
Über diesen Artikel
These torches light easily and stay lit, reducing downtime. With even and stable plasma to deliver reproducible results.
Frequently Bought Together
VWR®, Inserts for Short Thread Vials ND9
Anbieter: VWR Collection
VWR®, Funnels, Powder
Anbieter: VWR Collection
VWR®, Screw Neck Vials ND8
Anbieter: VWR Collection
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
VWR®, Screw Neck Vials ND24
Anbieter: VWR Collection
VWR®, Snap Cap Vials
Anbieter: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
VWR® Air Valve
Anbieter: VWR Collection
VWR®, Short Thread Vials ND9
Anbieter: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Anbieter: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Anbieter: VWR Collection
VWR®, Screw Neck Vials ND13
Anbieter: VWR Collection