Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ Florisil
Solid phase extraction cartridges, Bond Elut™ Florisil
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ Florisil
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ Florisil
15 Options Available Below
Product Details & Documents
Florisil is a magnesia-loaded silica gel, extremely polar in nature and ideal for the isolation of polar compounds from non-polar matrices.
  • Pesticide residue (PR) grade
  • Fast flow, ideal for viscous samples
The larger particle size of the sorbent enables flow for large sample volumes and is therefore an attractive alternative to silica if the sample matrix is particularly viscous.
Product Family Options
LRC cartridges
Product Information
  • Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12113049
    LRC cartridge
    LRC cartridge
    Florisil
    200 µm
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
Straight barrel cartridges
Product Information
  • Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12102024
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • VARI12102129
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • 551-0325
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • VARI12102109
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    3 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • VARI12102159
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
  • 551-0408
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
  • VARI52256014
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST
  • VARI12256022
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    12 ml
    2 g
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST
  • VARI12256047
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    20 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST
  • VARI12256046
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    20 ml
    2 g
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST
  • 551-0414
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    20 ml
    5 g
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST
  • VARI12256038
    Straight barrel cartridge
    Straight barrel cartridge
    Florisil
    200 µm
    60 ml
    10 g
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST
Bond Elut™ Jr
Product Information
  • Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12162050B
    Bond Elut™ Jr
    Bond Elut™ Jr
    Florisil
    200 µm
    500 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • VARI12166014B
    Bond Elut™ Jr
    Bond Elut™ Jr
    Florisil
    200 µm
    1000 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST