Fernbach culture flasks
Anbieter: SciLabware
Zertifikate
Über diesen Artikel
Culture flask, made of PYREX® borosilicate glass. For use in preparation of cultures.
- Exhibit 3 baffle indents to provide optimal and consistent agitation
- Large surface area to volume ratio
- Available in two versions, with Delong neck or screw cap
Frequently Bought Together