Culture flasks, narrow mouth, rimless
Supplier: SciLabware
Certificates
About this item
Culture flask, made up of PYREX® borosilicate glass with white enamel markings. Compatible with shaker equipment.
- Ideal for use in preparation of cultures
- Long, rimless tube neck for use with Aluminium or PP closures
- Excellent chemical and temperature resistance
Frequently Bought Together