Fluorescence cuvettes for flow-through meaurements
: HELLMA
The cuvettes are designed with a chamber to minimise bubble formation.
- Screw connections tested to 5 bar
- Cells with inlet/outlet tubes tested at 3 bar
- Contact surfaces treated to reduce bubble adhesion within the chamber
Suitable for high precision analytical techniques.
Frequently Bought Together