Condensers, coil, reversible
Nome del fornitore: SciLabware
Certificati
Informazioni su questo articolo
PYREX® borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Highly versatile reflux condensers. Each end of the condenser is fitted with a different size cone thus reducing the need for adapters. With detachable plastic connectors for the attachment of flexible tubing.
Frequently Bought Together