Volumetric flasks, class B, blue markings
Supplier: SciLabware
Jump to:
About this item
MBL®, borosilicate glass, with PE stopper.
- Marks and inscriptions in high contrast blue enamel
- Excellent temperature resistance
: Complies with ISO 1042 and DIN 12664
11 Options Available Below
Product Details & Documents
MBL®, borosilicate glass, with PE stopper.
- Marks and inscriptions in high contrast blue enamel
- Excellent temperature resistance
: Complies with ISO 1042 and DIN 12664
Product Family Options
Product Information
- CapacityNSHeightToleranceAvailabilityPrice