Orders LinkContactUsLinkComponent
Torion stopcocks, SLV®
Supplier:   SciLabware
Torion stopcocks, SLV®
Supplier:  SciLabware
Certificates
Torion stopcocks, SLV®
Torion stopcocks, SLV®
6 Options Available Below
Product Details & Documents
Stopcocks are made of borosilicate glass.
  • Vacuum capability to 10⁻² Torr
  • Sidearms are 70 mm in length
  • Excellent chemical and temperature resistance
In right angle pattern PTFE key is screwed into the threaded Torion connector, to open and close the stopcock.
Delivery information: Supplied with fittings, sealing rings and PTFE key.
Product Family Options
Right angle pattern
Product Information
  • Hole Ø
    For tube O-Ø
    Availability
    Price
  • 5,0 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 1’510.00
    SFr. 1’510.00
    1 * 10 ST
       SFr. 1’510.00
  • 8,7 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 1’730.00
    SFr. 1’730.00
    1 * 10 ST
       SFr. 1’730.00
  • 14,7 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 1’360.00
    SFr. 1’360.00
    1 * 5 ST
       SFr. 1’360.00
Straight pattern
Product Information
  • Hole Ø
    For tube O-Ø
    Availability
    Price
  • 5,0 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 988.00
    SFr. 988.00
    1 * 5 ST
       SFr. 988.00
  • 8,7 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 1’010.00
    SFr. 1’010.00
    1 * 5 ST
       SFr. 1’010.00
  • 14,7 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 1’540.00
    SFr. 1’540.00
    1 * 5 ST
       SFr. 1’540.00