Supelco® Organic Reference Standard, Jasmone
Katalognr. 59975-1ML
Anbieter: Merck
CAS-Nummer:
Technische Daten
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:488-10-8
- Description:Jasmone
- Storage conditions:2...30 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00001402
- M-Clarity quality level:MQ100
- Synonym(s):cis-3-Methyl-2-(2-pentenyl)-2-cyclopenten-1-one
- Molecular weight:164,24
- Density:0,94 g/ml at 25 °C (lit.)
- Flash point:107 °C - Closed cup
- Boiling point:134 - 135 °C/12 mmHg (lit.)
Technische Daten
Über diesen Artikel
- GHS signal: GHS07
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338