ContactUsLinkComponent
VWR® AVS TITRINORM, Bismuth Standard Solution, 1000 mg/L Bi in 10% Nitric Acid, Standard for AAS
VWR® AVS TITRINORM, Bismuth Standard Solution, 1000 mg/L Bi in 10% Nitric Acid, Standard for AAS
Supplier:  VWR Chemicals
Certificates
undefined
VWR® AVS TITRINORM, Bismuth Standard Solution, 1000 mg/L Bi in 10% Nitric Acid, Standard for AAS
Supplier:  VWR Chemicals