Centrifuge tubes, glass
PYREX®, borosilicate glass, cylindrical with round bottom.
- Uniform glass distribution in wall and base for improved strength
- Excellent chemical and thermal resistance
- Max. RCF 3000×g
Robust universal centrifuge tubes for many uses.
Frequently Bought Together