Joint clips for spherical joints, metal, Quickfit®
Anbieter: SciLabware
Zertifikate
Über diesen Artikel
Metal, corrosion resistant nickel finish. Clips in 'fork' type design for retaining glassware having Quickfit® spherical joints.
- Spring loaded to ensure a positive pressure is applied to both surfaces of the joint
- Suitable for high temperature work
Frequently Bought Together